Learn More
MilliporeSigma™ Supelco™ Glass Packed GC Column (fits Agilent, Configuration A), Phase: 10 OV-101, Matrix 80/100 Chromosorb W HP Support
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness.
Manufacturer:  MilliporeSigma™ Supelco™ 13086U
Catalog No. 11-100-9500
Add to cart
Description
Description
This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
Specifications
Specifications
Gas Chromatography (GC)
Packed GC
Glass Column
2 mm
0.25 in.
1.8 m
OV-101, Matrix 80/100 Chromosorb W HP Support