Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Glass Jointed Powder Funnels
Ideal for transferring powder reagents between pieces of labware. Eisco™ Glass Jointed Powder Funnels feature ground joints to ensure tight connections with other pieces of glassware.
Supplier: Eisco™ CH0460D
Description
- Made from high quality borosilicate 3.3 glass
- Designed with ground joints to ensure tight connections with other pieces of glassware
- Ideal for transferring powder reagents
- 60° angle stem
- Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
70 mm | |
12.7 cm | |
Autoclavable | |
No | |
29/32 | |
5 in. |
Borosilicate Glass | |
60° | |
Clear | |
Transferring liquids | |
1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction