Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Medium Porosity, Tube O.D./13mm
Supplier: Chemglass Life Sciences CG20202
Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Medium Porosity, Tube O.D./13mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.