Learn More
Polysciences, Inc. GLASS BEADS 50-100UM 100GM / specially selected soda lime glass with a density of ~2.4
Supplier: Polysciences, Inc. 15926100
GLASS BEADS 50-100UM 100GM / specially selected soda lime glass with a density of ~2.4