Learn More
Polysciences, Inc. GLASS BEADS 105-150UM 100GM / specially selected soda lime glass with a density of ~2.4
Supplier: Polysciences, Inc. 15927100
GLASS BEADS 105-150UM 100GM / specially selected soda lime glass with a density of ~2.4