Learn More
Gibco™ Fetal Bovine Serum, qualified, New Zealand
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Supplier: Gibco™ A3160902
Description
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)
- Bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label has all the information you need
- Generates 33% less waste than aliquoting from 500mL bottles
- Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Premium (Secure)
- Origin: New Zealand
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Bovine | |
One Shot | |
10 x 50 mL | |
Sterile | |
Fewer resources used and less waste | |
Qualified | |
Frozen |
Fetal Bovine Serum, qualified, One Shot™ format, New Zealand | |
New Zealand | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
≤30 mg/dL | |
One Shot™ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For In Vitro Diagnostic Use.