Learn More
COLUMBIA ANALYTICAL INSTRUMENTS INC GGA1X APHA STANDARD 300 BOX
Supplier: COLUMBIA ANALYTICAL INSTRUMENTS INC GGA1X.300
Box of 300 Single-strength GGA Standard for BOD5 compliance testing. 30 month shelf life means the most stable product avaialble! QC Document included.