Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Germaine™ Laboratories Febrile Test Controls
For use as positive and negative control for Febrile antigen tests
$64.50 - $82.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
16-414-114
|
Germaine™ Laboratories
1600 |
Febrile Positive Control |
Each for $82.00
|
|
|||||
16-414-107
|
Germaine™ Laboratories
1601 |
Febrile Negative Control |
Each for $64.50
|
|
|||||
Description
- 5mL vial
Specifications
5 mL |
Professional use only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title