Learn More
Applied Biosystems™ GeneScan™ 350 TAMRA™ dye Size Standard
GeneScan 350 TAMRA Size Standard is designed for sizing DNA fragment in the 35-350 nucleotides range and provides 12 single-stranded labeled fragment.
Manufacturer:  Applied Biosystems™ 401736
Encompass
Catalog No. 40-173-6
Add to cart
Includes: Contains12 TAMRA labeled single stranded DNA fragments for use as a standard
Description
Description
The GeneScan™ 500 XL TAMRA™ Size Standard is a TAMRA™ dye-labeled size standard for the reproducible sizing of fragment analysis data.
Order Info
Each
Specifications
Specifications
Contains12 TAMRA labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze).
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
TAMRA™
GeneScan™, TAMRA™
Wet Ice
Standard
Sequencing
800 Reactions
Dye
Standard