Learn More
Andwin Scientific GENERAL PURPOSE MICROWAVE OVEN
SDP
Supplier:  Andwin Scientific PX17088
Encompass_Preferred
The Panasonic Genius 2.2 cu. ft full-size inverter microwave has a 5-button control panel with an added dial for temperature control and a keep warm function to maintain temperatures at a constant level without overcooking. This microwave also features a menu action screen, one-touch genius sensor, and a versatile function key that simplifies the programming of 10 useful non-cooking functions. This product has a 1200 W output power.
Catalog No. NC9395948
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.