Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers Safety General-Purpose 6 in. Non-Lighted Warning Whip w/ Threaded Hex Base 12 in. Orange Flag White X

Includes Day Bright™ mesh flag with 3M™ Scotchlite™ reflective X, fiberglass resin whip, base mount

Supplier:  Checkers Safety FS6XO

Catalog No. 19-121-122

This item has been discontinued and is no longer available. Please reach out to your sales representative.


Description

Description

  • Threaded hex base warning whip length: 6 ft.
  • Flag color: orange
  • Includes:
    • Heavy duty Day Bright™ fluorescent nylon mesh flag with 3M™ Scotchlite™ reflective X
    • Premium fiberglass resin whip
    • Choice of base mount
  • Specifications

    Specifications

    Non-lighted general purpose safety warning whip
    Threaded Hex base mount
    6 ft.
    Orange
    12 x 12 in. (flag)
    Warning Whip
    Nylon Mesh, Fiberglass Resin
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit