Learn More
Thermo Scientific GD AS18 FAST 4X150MM
Supplier: Thermo Scientific 072063
For Use With IonPac AS18-fast Capillary Column. •Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A) •Determination of low levels of inorganic anions in complex matrices using large-loop injections •Superior retention and quantification of fluoride, acetate, and formate