Learn More
Andwin Scientific GAS LIGHTER
Supplier: Andwin Scientific LX3574
Piezo Electric Gas Lighter, Small electric generator produces sparks at 20Kv to provide safe flameless ignition of gas burners and torches. Plastic pistol-grip, aluminum barrel and button trigger to accurately aim spark. Length: 26.5cm.