Learn More
Diversified Spaces™ Gas Cocks
Choose from three styles of Diversified Gas Cocks for your laboratory needs.
Supplier: Diversified Spaces™ 100047
This item is not returnable.
View return policy
Description
Units complete with nipple, locknut and washer for installation. 90, 180 and 360° models have two or four nipples at said angle.
- Chrome-plated brass
- Deck-mounted
- Ground key valve for long life
Specifications
Chrome-plated Brass | |
Nipple, locknut and washer, deck mounted, ground key valve | |
5 cm | |
17.78 cm | |
12.7 cm | |
0.9 kg |
Gas Cock | |
2 in. | |
7 in. | |
5 in. | |
2 lb. |