Learn More
CROSSTEX INTERNATIONAL Indicator Labels for hand-held labeling systems, 25 mm x 12 mm, 1000/roll
Supplier: CROSSTEX INTERNATIONAL CPIR02
Intended for use with individual units (e.g. packs, containers) to indicate that the unit has been directly exposed to a Gamma or E-Beam process and to distinguish between processed and unprocessed units.