Learn More
Chemglass Life Sciences 60mL Addition Funnel, Graduated, 14/20 Joint Size, 0-2mm PTFE Valve, 235mm OAH
Supplier: Chemglass Life Sciences CG171412
60mL Addition Funnel, Graduated, 14/20 Joint Size, 0-2mm PTFE Valve, 235mm OAH. Cylindrical style graduated addition funnel has a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.