Learn More
IDEX Health & Science Upchurch Stainless Steel Frit 0.5µm .062" x .065" x .2485
Supplier: IDEX Health & Science Upchurch A102
Our Stainless Steel Frit is designed specifically to work well throughout HPLC systems. We design these frits to deliver excellent uniform porosity and a long filtration life. It has a porosity of .5µm.