Learn More
Foss™ Digestion System: Scrubber Units: SR210, automatic version
For neutralization of corrosive fumes in acid digestions
Supplier: Foss™ 60063222
Description
Available manual or automatic and programmable.
- Can be combined with any of the exhaust manifolds
- Handles up to 100 samples without changing of reagent
- Compact benchtop unit is self-contained and therefore unaffected by water supply issues
- When connected to Auto Lift or Auto Rack system, the auto program with fully control function including switching from high to low aspiration settings
- TC2501001: automatic and programmable
- 12053110: not automatic or programmable
Specifications
Scrubber Unit | |
110/230 V | |
Fully automated and programmable neutralization of corrosive fumes in acid digestions | |
1 Ea. |
50/60 Hz | |
110/230 V, 50/60 Hz | |
Acid Digester |