Learn More
Sigma Aldrich Fine Chemicals Biosciences Formamide, 75-12-7, MFCD00007941, 100 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences F9037100ML
Linear Formula: HCONH2, Molecular Weight: 45.04, Synonym: Amide C1, Formic amide. ≥99.5% (GC), BioReagent, for molecular biology. Formamide is a clear liquid amide derived from formic acid. It can be used as a cryoprotectant and gel-stabilizer. It is known to destabilize nucleic acid duplexes, making it useful for preparing hybridization buffers for RNA and DNA applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.