Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v) Neutralized Solution, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
50-00-0, 7732-18-5, 67-56-1
50-00-0, 7732-18-5, 67-56-1
Packaging Poly Bottle
Quantity 500 mL
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.