Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formalde-Fresh Solution, Fisher Chemical™
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Physical Form | Liquid |
Melting Point | 0°C |
Boiling Point | 93.9°C |
Density | 1.10g/cm³ |
Packaging | Poly Bottle |
pH | 6.9 to 7.1 |
Quantity | 4 L |
Assay Percent Range | 3.5 to 4.5 % |
Percent Purity | 3.5 to 4.5% |
Chemical Name or Material | Formalde-Fresh Solution |
Safety and Handling
Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Cancer hazard. May cause an allergic skin reaction. Irritating to eyes, respiratory system and skin. May be harmful by inhalation, in contact with skin and if swallowed. May cause central nervous system effects. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Take precautionary measures against static discharges. Do not get in eyes, on skin, or on clothing. Do not breathe vapors or spray mist. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:2 Flammability:2 Instability:0 |
DOTInformation | DOT Class 9: Miscellaneous |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction