Learn More
IMEB, Inc Pathology Bone Band Saw Optional Foot Pedal Switch
Supplier: IMEB, Inc FS351
Foot pedal switch for IMEB bone band saw. IMEB Bone Band Saw sold separately. The bone band saw only is 19″ high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.