Learn More
Applied Biosystems™ Fluorescent Amidite Matrix Standards [6FAM™, TET, HEX, TAMRA™, and ROX™ dyes]
The ExoSAP-IT one-step solution provides a simple way to enzymatically purify a PCR reaction.
Manufacturer: Applied Biosystems™ 401546
Includes: 5 Tubes of dye labeled oligos, one tube for each dye
Description
The Fluorescent Amidite Matrix Standards are used to generate the multicomponent matrix required when analyzing 6-FAM™, TET™, HEX™, TAMRA™ and ROX™-labeled DNA fragments.Order Info
Each
Specifications
Contains 5 tubes of dye labeled oligos, one tube for each dye. Store the kit at 2-8°C. Do not freeze. | |
Sequencing | |
FAM™, HEX™, ROX™, TAMRA™, TET™ | |
1 Each |
310 Genetic Analyzer | |
FAM, HEX™, ROX, TAMRA™, TET™ | |
DNA Sequencing Reagents | |
Wet Ice |