Learn More
Polysciences, Inc. Fluoresbrite YG Microspheres 1.00um
SDP
Supplier:  Polysciences, Inc. 1715410
Encompass_Preferred
Fluoresbrite Yellow Green Microspheres have excitation and emission spectra similar to FITC with excitation maxima of 441nm and emission maxima at 485nm. They are one of the brightest microspheres available at this wavelength and are used extensively in phagocytosis studies, flow cytometry and diagnostic assays. Nominal Reference Values: 1.00m particles packaged as 2.5% aqueous suspension, 4.55 x 1010 particles/ml, Coefficient of Variation (CV) = 3%, Excitation max. = 441nm, Emission max. = 486nm, Storage: Store at 4 degrees celsius; Do not permit to freeze.
Catalog No. NC9643434