Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorene, 98%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
86-73-7
86-73-7
CAS Max % 1
Packaging Poly Bottle
Quantity 25 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.