Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Fluka™ QuEChERS, Sample Extraction
Extraction Salt for QuEChERS Method with 50 mL Tube
Catalog Number | Mfr. No. | Sorbent | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sorbent | Quantity | Price | Quantity & Availability | ||||
600-29-66
|
Honeywell™
5601 |
4 gm MgSO4, 1 gm sodium chloride per Tube | 50 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-29-68
|
Honeywell™
5627 |
4 gm MgSO4, 1 gm sodium chloride per Tube | 250 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-29-69
|
Honeywell™
5602 |
4 gm MgSO4, 1 gm sodium chloride, 500 mg sodium citrate (dibasic sesquihydrate),1 gm sodium citrate (tribasic dihydrate) per Tube | 50 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-29-72
|
Honeywell™
5626 |
6 gm MgSO4, 1.5 gm sodium acetate per Tube | 250 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
QuEChERS
- Quick, Easy, Cheap, Effective, Rugged and Safe Sample Extraction and Cleanup Products
- Honeywell Burdick & Jackson™ QuEChERS products offer an alternative sample preparation technique to traditional liquid-liquid and solid phase extraction (SPE) methods
- the use of QuEChERS has the potential to lower the overall cost of sample preparation due to the increased efficiency and ease-of use when compared with SPE methods
Specifications
50 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title