Learn More
Chemglass Life Sciences 50mL Flask, Reaction, 14/20 Outer Joint, 2mm Glass Stopcock, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF052003
50mL Flask, Reaction, 14/20 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids. Lower portion of flask fits standard heating mantles. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.