Learn More
R&D Systems™ FlowX Human Th2 Cell Multi-Color Flow Cytometry Kit
R&D Systems™ FlowX Human Th2 Cell Multi-Color Flow Cytometry Kit is designed for the flow cytometric analysis of Th2 cells using five fluorochrome-conjugated antibodies.
Supplier:  R&D Systems™ FMC011B
Catalog No. FMC011B
Add to Cart
This item is not returnable. View return policy
Description
Description
  • This kit contains five conjugated antibodies and buffers that can be used for staining of human Th2 cells (1-6)
  • Positive Markers:
    • 125μL of CD4 Alexa Fluor™ 700 Mouse IgG2A (Clone 11830)
    • 125μL of IL-10 Alexa Fluor™ 488 Rabbit IgG (Clone 2050B)
    • 250μL of CCR3-PE Rat IgG2A (Clone 61828)
    • 125μL of GATA-3 Alexa Fluor™ 594 Mouse IgG2B (Clone 634919)
    • 125μL of Phospho-STAT6 Alexa Fluor™ 647 Rabbit IgG (Clone 1248D)
  • Fixation/Permeabilization Buffer (1X):
    • 30mL of 1X fixation buffer.
  • Staining Buffer (1X):
    • 50mL of a 1X staining buffer
  • Some components in this kit contain formaldehyde which is a suspected carcinogen.
  • Avoid contact with skin, eyes, and mucous membranes, and avoid inhaling fumes.
  • In case of contact, wash immediately with water and seek medical advice.
  • Some components in this kit contain sodium azide which may react with lead and copper plumbing to form explosive metallic azides.
  • Flush with large volumes of water during disposal.
Specifications
Specifications
25
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Suggestions
Product Suggestions