Learn More
R&D Systems™ FlowX Human Regulatory T Cell Multi-Color Flow Kit
For the single-step staining of human regulatory T cells (1-9)
Supplier:  R&D Systems™ FMC021
Catalog No. FMC021
Add to Cart
This item is not returnable. View return policy
Description
Description
Contains conjugated antibodies to CD25-APC (Clone 24212), FoxP3-PE (Clone 1054C), CD4-CFS (Clone 11830)
Specifications
Specifications
Multi-Color Flow Kit
25 Tests
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Suggestions
Product Suggestions