Learn More
Perkin Elmer US LLC Flow Spoiler for Legacy AA Instruments
Supplier: Perkin Elmer US LLC 00572561
This flow spoiler is installed in the burner chamber and is compatible with PerkinElmer atomic absorption flame instruments Models 2280, 2380, 3030(B), 3100, 3110, 3300, 4000, 5000, and 5100 PC.