Learn More
Chemglass Life Sciences Flow Indicator, Polystyrene
Supplier: Chemglass Life Sciences CG160501
Flow Indicator, Polystyrene. Clear flow indicator provides immediate visual of water, air or gas flow. The highly visible white nylon ball rotates in a circular channel during flow. Continuity and relative flow rate are indicated by the ball’s rotation. The minimum flow rate to actuate the ball with water is 50mL per minute in the horizontal position, 140mL per minute in the vertical position. With air, the minimum flow rate is 3L/min in the horizontal position, 8L/min in the vertical position. Maximum working pressure is 10 PSI. Hose connections accept 1/4 to 3/8" ID tubing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.