Learn More
Vector Laboratories Fluorescein Avidin D (FITC) 5 mg
Supplier: Vector Laboratories A20015
Fluorescein Avidin D is made from highly purified Avidin D and has an optimal fluorochrome to protein ratio. Compared to conventional primary and secondary fluorescent labeling techniques, Fluorescein Avidin D can provide greater sensitivity and lower background staining. For even higher sensitivity and specific applications, see Fluorescein Avidin DCS. Fluorescein Avidin D has an excitation at 495 nm and emission at 515 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.