Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Florisil Cartridge Quality Control Standard Mixture, Restek™
Click to view available options
Quantity:
10 mL
Description
- Reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography flame ionization detection (GC-FID) method.
- For GC analysis of total petroleum hydrocarbons (TPH) in water.
- Environmentally safer
- Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil
- Components:
- Diesel #2 (additive-free)
- Mineral oil (additive-free [i.e., USP grade]
Specifications
Concentration | 1,000 μg/mL each in cyclohexane, Total hydrocarbon concentration 2,000 μg/mL |
Packaging | Ampule |
Quantity | 10 mL |
Type | Multi-component Standard |
Safety and Handling
ShelfLife | 6 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction