Learn More
Essendant Stride Neutral Cleaner, Citrus Scent, 1.4 mL, 2 Bottles/Carton
Supplier:  Essendant DVO95122613
Encompass_Preferred
Concentrated formula for everyday cleaning of floors and other hard surfaces. Non-alkaline formula does not dull floor finishes. Versatile design to be used in automatic scrubbers, mop buckets and spray bottles.
Catalog No. NC1554323