Learn More
Bartels™ FlexTrans™ Transport Medium
The Bartels™ FlexTrans™ Transport Medium is intended to stabilize viruses and chlaymdiae, and suppress microbial contamination during transport of clinical specimens from the point of collection to the testing site. For in vitro Diagnostic Use. Store at room temperature, 2-8°C or -70°C
Supplier:  Bartels™ B102990D
Encompass
Catalog No. 22-140-361
Add to Cart
This item is not returnable. View return policy
Description
Description
FlexTrans™ - Transport medium in a 15 mL conical centrifuge tube, allowing specimen collection, transport and processing in the same container. Medium contains Minimal Essential Medium supplemented with L-glutamine and Hanks Salts, bovine serum albumin, sucrose, amphotericin B, gentamicin and streptomycin buffered with HEPES Buffer to a pH range of 7.0-7.4. Phenol red is added as a pH indicator. The transport tube also contains glass beads to aid in the disruption of patient cells in the specimen, with subsequent release of viruses or chlamydiae into the medium.
Specifications
Specifications
3 mL of transport medium in a 15 mL conical centrifuge tube
50 Tubes/Pk.
Store at room temperature, 2°C to 8°C or -70°C
Product Suggestions
Product Suggestions