Learn More
Chemglass Life Sciences 20L Flask, Evaporating, Large Scale, 150mm Flange, 425mm OAH
Supplier: Chemglass Life Sciences CG133709
20L Flask, Evaporating, Large Scale, 150mm Flange, 425mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.