Learn More
Chemglass Life Sciences 10L Flask, Evaporating, Large Scale, 150mm Flange, 415mm OAH
Supplier: Chemglass Life Sciences CG133708
10L Flask, Evaporating, Large Scale, 150mm Flange, 415mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.