Learn More
Structure Probe Inc FLASH-DRY SILVER PAINT 1/2 TR
Supplier: Structure Probe Inc 04998AB
SPI Flash-Dry Ag Colloidal Suspension with Brush Applicator Cap, 0.5 troy oz.(15.5g), (OK-SPI) Flammable Material