Learn More
MilliporeSigma™ Supelco™ Flash-Chromatography Assembly with Standard Column and Threaded Joints , Joint Size: ST/NS 45/50
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$868.00 - $911.00
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size ST/NS 45/50
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
11-100-7814
View Documents
MilliporeSigma™ Supelco™
Z1473971EA
1000 mL 57 mm 508 mm
Each for $868.00
Add to cart
11-100-7721
View Documents
MilliporeSigma™ Supelco™
Z1474001EA
2000 mL 76 mm 560 mm
Each for $911.00
Add to cart
Description
Description
Grease-free design eliminates contamination and frozen joints. Consists of column and flow controller. Order solvent reservoir separately.
Specifications
Specifications
Flash Chromatography
Flash Chromatography
ST/NS 45/50