Learn More
MilliporeSigma™ Supelco™ Flash Chromatography Assembly with Standard Column/Ball Joints, Joint Size: SJ 51/30
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z2023981EA
 View more versions of this product
Catalog No. 11-100-8083
Add to cart
Description
Description
Consists of column, flow controller, and pinch clamp. Order solvent reservoir separately.
Specifications
Specifications
560 mm
Flash Chromatography
400 mL
38 mm
Flash Chromatography
SJ 51/30