Learn More
Haztech Systems, Inc Hazcat Flame Test Wires
Supplier: Haztech Systems, Inc HW4024
Hazcat's Flame Test Wires are perfect for detecting metals. Able to detect the following metals: Copper, sodium, potassium, lithium, calcium, molybdenum, strontium, thallium, tellurium, manganese, lead, selenium, zinc, rubidium, cesium and more. Ni Cd wire, Noncalibrated 2mm 24 ga. loop /12ct.
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'