Learn More
Fisherbrand™ White Line Flasks with Standard Taper Stoppers
Single graduation ring blasted on the neck.
$415.00 - $490.00
Specifications
Calibration | Calibrated To Contain |
---|---|
Closure Material | Glass |
Material | Borosilicate Glass |
No. per Case | 6 |
Graduated | Yes |
Includes: 41900R ground glass stopper with marking spot
Description
- Calibrated “To Contain”
- Reusable
- Class A volumetric
Compliance
Designed from ASTM™ Specification E 288, Class A unserialized requirements
Specifications
Calibrated To Contain | |
Borosilicate Glass | |
Yes |
Glass | |
6 |