Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ SQ-EASY™ PreFilter Tips
Disposable, efficient self-contained filtration system for sample preparation applications.
Specifications
Product Type | Sq-Easy™ Filter Tip |
---|---|
For Use With (Application) | For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays |
For Use With (Equipment) | PreFilter™ system |
Quantity | 100 Pack |
Tip Type | Hydrophilic filter |
Catalog Number | Mfr. No. | Material | Particle Retention | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Particle Retention | Quantity | Price | Quantity & Availability | |||
22-267-389
|
Fisherbrand™
X79840 |
HDPE | 15μm | 100 Pack | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||
Description
- For use in stool, serum, diluted whole blood, urine, and throat swab assays
- Components are liquid-tight and interlocking to prevent sample leakage
- Range of filtration materials available, including ultra-high molecular weight polyethylene (UHMWPE), high-density polyethylene (HDPE), and glass fiber
- Reagents can be impregnated directly into the filter materials
- All components are available separately to alllow for fllexibility in tube size and particle retention
Specifications
Sq-Easy™ Filter Tip | |
PreFilter™ system | |
Hydrophilic filter |
For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays | |
100 Pack |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title