Learn More
Fisherbrand™ Natural Color Micro Pipet Tips
For use with micro volume pipetters
Description
- Natural color
- S66803 is packaged 1000 tips per pack; others have 960 tips
Specifications
0.1 to 10 μL | |
Natural | |
3.18cm | |
MicroPoint | |
Polypropylene |
Non-sterile | |
1.25 in. | |
Hinged Rack | |
Universal Fit |