Learn More
Fisherbrand™ Gas Cylinder Support
Keeps cylinders upright and safe.
This item is not returnable.
View return policy
Description
- Nylon strap securely holds 4.5 to 11 in. diameter (11.4 to 28cm) cylinders upright
- Can be attached to benches up to 2.25 in. thick (5.7cm), with at least 1 in. (2.5cm) overhang
Specifications
4.5 to 11 in. dia. cylinders upright | |
47 in. long nylon strap | |
Gas Cylinder Support | |
Gas cylinder |