Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
This item is not returnable.
View return policy
Description
- Use in any chemical environment
- Withstand temperatures up to 1150°C (2,102°F)
- Consistent wall thickness and density
- Fired twice to give added strength and durability
- Highly resistant to chemical attack
Order Info
Crucible covers sold separately (Cat. Nos. S02232 through -238).
Specifications
Porcelain | |
Used in chemical environments | |
250 mL |
Smooth, Glazed | |
1150°C | |
Wide-form |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction