Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ NMR PTFE Sample Tube Pressure Cap for 5mm Tube

White PTFE Caps

Catalog No. K8970960003


Add to cart

Description

Description

  • Fits 5mm tube size
  • Pack of 25

Specifications

Specifications

Not Autoclavable
White
No
Non-sterile
3 mm Standard or Medallion Series NMR Sample Tube
PTFE
3 mm
Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
Pressure Cap
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit