Learn More
Fisherbrand™ Isotemp™ Water Baths
Advanced features offer improved accuracy, safety and ease of use.
This item is not returnable.
View return policy
Description
Deep drawn stainless-steel reservoir and perforated sample tray eliminates leaks and guards against rust and contamination. Digital temperature set and readout let you easily program the exact temperature you want between ambient and 100°C.
- Provide ±0.24°C uniformity and ±0.5°C temperature stability at 37°C (98.6°F)
- Easy-to-read display is selectable for °C or °F
- Temperature-resistant plastic rim improves resistance to leakage from water runoff
- Hinged polycarbonate gable covers stay open at 90° position or lift off completely using quick-release hinge to accommodate large glassware
- Dual thermostats safeguard against thermal runaway
- Heating elements will not burn out if bath accidentally runs dry
- Cover extension fins keep hands away from hot vapors
- Outer plastic casing remains cool during operation
Specifications
5 L | |
Polycarbonate gable cover | |
5.88 x 11.75 x 2.5” | |
10.5 in. | |
14.1 in. | |
7.63 in. |
100°C | |
±0.5°C | |
10.5 x 14.1 x 7.63 in. (26.7 x 35.8 x 19.4 cm) | |
26.7 cm | |
35.8 cm | |
19.4 cm |