Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Flammable-Materials Storage Refrigerators: Basic Model

For reliable, safe storage of volatile materials, pharmaceuticals, clinical reagents, and biological samples

 View more versions of this product

Catalog No. 50FREEFSA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

  • Adjustable temperatures from 1° to 12°C, depending on model
  • 50 cu. ft. (1420L) model features digital temperature display in 0.1° increments, °C or °F, and keyed power switches
  • Manual defrost
Safety Features
  • Orange warning label on front
  • Sparkproof interiors separate contents from compressor and motor
  • Compressors have enclosed overloads, relays, and thermostats that prevent sparks from escaping during door opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Top-mounted hydraulic thermostat and door stop
Construction
  • White, baked-on polyester enamel exteriors CFC-free polyurethane foam insulation and refrigerant
  • Solid door(s)

Service Plan

Service agreement available.

Certifications

UL listed meets NFPA 45 and 70

Compliance

NFPA Code 99 (30 and 50 cu.ft. models only)

Warranty and Services

13 months, parts and labor

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flammable Material Storage Refrigerator
1420 L
R134a, 100% CFC Free
2 (2 compartments)
Enameled Steel
Stainless Steel
4 Levelling Legs
Mechanical Thermostat/ Outside of Unit, Top Mounted
Manual
Door Lock
26.5 x 45.75 x 59 in. (67.3 x 116.2 x 149.8 cm)
5 A
120 V
60 Hz
50 cu. ft.
1°C to 12°C
Hinged, Open Outwards
8 Adjustable
Polyurethane Foam
UL
Gray
13 Months Parts and Labor
50FREEFSA
Top-mounted
34.25 x 52 x 77.25 in. (87 x 132.1 x 196.2 cm)
NEMA 5-15 P
115 V 60 Hz
600 w
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit