Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Isotemp™ Economy Analog-Control Water Baths
Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical and biomedical procedures
Specifications
Controller Type | Analog |
---|---|
Certifications/Compliance | CSA |
Temperature Range | 5°C to 75/100°C |
For Use With (Application) | Industrial, clinical, pharmaceutical or biomedical procedures |
Includes | Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug |
Includes: Sample tray; Thermometer clip; Nonhazardous liquid, partial-immersion (76mm) thermometer (-10° to +110°C); Line cord and grounded NEMA 5-15P plug
Catalog Number | Dimensions (L x W x H) | Model | Shipping Weight (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Dimensions (L x W x H) | Model | Shipping Weight (English) | Price | Quantity & Availability | ||||
15-460-20Q
|
16.53 x 24.38 x 8.88 in. (42 x 62 x 23 cm) | 120 | 22.25 lb. |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
15-460-5Q
|
10.5 x 14.17 x 7.48 in. (27 x 36 x 19 cm) | 105 | 9.88 lb. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Easy-to-use controls on front panel for full access and safe operation
- Dual thermostats guard against thermal runaway; elements won't burn out even if bath runs dry
- Choice of acrylic or stainless-steel covers
- Hinged stainless-steel or see-through acrylic gable covers have unique fins that keep hands away from hot vapors; stay open at 90° position or lift off to accommodate large glassware
- Corrosion-resistant exterior with acrylic enamel finish; temperature-resistant plastic rim fits snugly to insulate and prevent leaks
- Outside of the bath stays cool to the touch even after hours of operation
- Low-profile design makes access convenient
- Deep-drawn stainless-steel reservoirs are seamless to prevent leaks; rounded corners make cleaning easy
- Shallow-form Model 102S is perfect for heating samples in microcentrifuge tubes and for other molecular biology procedures
- Model 115 has two chambers, each with its own temperature control panel
Order Info
Concentric ring covers and stainless-steel gable covers are available separately.
Service Plan
Service agreement available.
Certifications
CSA approved.
Warranty and Services
One-year warranty on parts, labor and travel.
Specifications
Analog | |
5°C to 75/100°C | |
Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug | |
1 Year | |
50/60 Hz |
CSA | |
Industrial, clinical, pharmaceutical or biomedical procedures | |
±0.2°C | |
120 V |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title